Ethyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylate 98+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,5,6,7-tetrahydrobenzo[b]thiophene-2-carboxylate

A115799-1G A115799-250MG A115799-5G
77009-154EA 35 USD
77009-154 77889-793 77889-794
Ethyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylate 98+%
Ethyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂S
MDL Number: MFCD06662278
CAS Number: 19282-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR