trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: trans-4'-Propyl-(1,1'-bicyclohexyl)-4-carboxylic acid

A115661-100G A115661-10G A115661-1G A115661-25G A115661-5G
77026-072EA 108.22 USD
77026-072 77026-074 77026-076 77026-078 77026-080
trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid 97%
trans,trans-4'-n-Propylbicyclohexyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₈O₂
MW: 252.398 g/mol
MDL Number: MFCD05864407
CAS Number: 65355-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR