4,6-Dichloro-2,3-dimethylpyridine 97%

Supplier: Ambeed
Warning

A115584-100MG A115584-250MG
76982-946EA 97.61 USD
76982-946 76982-948
4,6-Dichloro-2,3-dimethylpyridine 97%
4,6-Dichloro-2,3-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
MDL Number: MFCD26743449
CAS Number: 101252-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR