Ethyl 5-aminothiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A115563-100MG A115563-1G A115563-250MG A115563-5G
77026-064EA 8.49 USD
77026-064 77026-066 77026-068 77026-070
Ethyl 5-aminothiazole-4-carboxylate 97%
Ethyl 5-aminothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
MDL Number: MFCD09834584
CAS Number: 18903-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR