3-Iodophenyl acetate 95%

Supplier: Ambeed
Warning

A115515-1G A115515-250MG
76982-942EA 33.95 USD
76982-942 76982-944
3-Iodophenyl acetate 95%
3-Iodophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.04 g/mol
MDL Number: MFCD09029734
CAS Number: 42861-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR