Isopentyl-4-(dimethylamino)benzoate 98%

Supplier: Ambeed
Warning

A115445-10G A115445-25G
77026-054EA 10 USD
77026-054 77026-056
Isopentyl-4-(dimethylamino)benzoate 98%
Isopentyl-4-(dimethylamino)benzoate

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD00059356
CAS Number: 21245-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR