9-Bromo-1-nonene 98%

Supplier: Ambeed
Warning

A115079-100G A115079-1G A115079-25G A115079-5G
77026-032EA 319.34 USD
77026-032 77026-034 77026-036 77026-038
9-Bromo-1-nonene 98%
9-Bromo-1-nonene

Precaución: for Research Use Only

Formula: C₉H₁₇Br
MW: 205.139 g/mol
MDL Number: MFCD09037826
CAS Number: 89359-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR