Ethyl 6-fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

A114840-100MG A114840-1G A114840-250MG A114840-5G
77026-018EA 5.31 USD
77026-018 77026-020 77026-022 77026-024
Ethyl 6-fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylate 95%
Ethyl 6-fluoro-1-methyl-4-oxo-7-(piperazin-1-yl)-1,4-dihydro-[1,3]thiazeto[3,2-a]quinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₀FN₃O₃S
MW: 377.43 g/mol
MDL Number: MFCD08063938
CAS Number: 113028-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR