4-Fluoro-2-hydroxybenzonitrile 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A114785-1G A114785-5G A114785-25G A114785-250MG
76982-914EA 27 USD
76982-914 77566-034 77610-924 77683-047
4-Fluoro-2-hydroxybenzonitrile 98%, Ambeed.Inc
4-Fluoro-2-hydroxybenzonitrile
Formula: C₇H₄FNO
MDL Number: MFCD13185487
CAS Number: 186590-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR