2-Bromo-3-methoxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A114725-100MG
A114725-250MG
A114725-1G
A114725-10G
A114725-5G
77009-120EA
37.14
USD
77009-120
77009-122
77566-028
77610-922
77683-046
2-Bromo-3-methoxybenzaldehyde 97%
2-Bromo-3-methoxybenzaldehyde
Precaución: For research use only.
Formula:
C₈H₇BrO₂ MW: 215.04 g/mol |
MDL Number:
MFCD09834663 CAS Number: 10401-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...