6-Chloro-1-indanone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2,3-dihydroinden-1-one

A114698-1G A114698-250MG A114698-5G
76995-448EA 9.55 USD
76995-448 76995-450 76995-452
6-Chloro-1-indanone 98%
6-Chloro-1-indanone

Precaución: for Research Use Only

Formula: C₉H₇ClO
MW: 166.61 g/mol
MDL Number: MFCD00956388
CAS Number: 14548-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR