4-Bromo-3-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Bromo-3-methylphenyl)methanol

A114685-10G A114685-1G A114685-5G A114685-250MG
77026-008EA 169 USD
77026-008 77026-010 77026-012 77871-473
4-Bromo-3-methylbenzyl alcohol 97%
4-Bromo-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD09264005
CAS Number: 149104-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR