2-Amino-3-(o-tolyl)propanoic acid 97%

Supplier: Ambeed
Warning

A114650-10G A114650-250MG A114650-5G
76995-438EA 285 USD
76995-438 76995-440 76995-442
2-Amino-3-(o-tolyl)propanoic acid 97%
2-Amino-3-(o-tolyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD01823323
CAS Number: 22888-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR