2,4-Difluoro-D-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-3-(2,4-difluorophenyl)propanoic acid

A114599-1G A114599-250MG
76995-434EA 70.02 USD
76995-434 76995-436
2,4-Difluoro-D-phenylalanine 98%
2,4-Difluoro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
MDL Number: MFCD07371994
CAS Number: 266360-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR