1-(4-Hydroxy-5-methoxy-2-nitrophenyl)ethanone 97%

Supplier: Ambeed
Warning

A114575-100MG A114575-1G A114575-250MG
77009-110EA 38 USD
77009-110 77009-112 77009-114
1-(4-Hydroxy-5-methoxy-2-nitrophenyl)ethanone 97%
1-(4-Hydroxy-5-methoxy-2-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MW: 211.17 g/mol
MDL Number: MFCD27996347
CAS Number: 418759-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR