GSK126 ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (S)-1-(sec-Butyl)-N-((4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-3-methyl-6-(6-(piperazin-1-yl)pyridin-3-yl)-1H-indole-4-carboxamide

A114568-10MG A114568-1MG A114568-25MG A114568-5MG A114568-100MG A114568-250MG A114568-50MG
76982-906EA 46.68 USD
76982-906 76982-908 76982-910 76982-912 77683-041 77683-042 77683-043
GSK126 ≥98%, Ambeed.Inc
GSK126

Precaución: For research use only.

Formula: C₃₁H₃₈N₆O₂
MW: 526.67 g/mol
MDL Number: MFCD23381067
CAS Number: 1346574-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR