Methyl-3-oxocyclobutanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-oxocyclobutanecarboxylate

A114502-100G A114502-10G A114502-25G A114502-500G A114502-5G
77025-986EA 126.25 USD
77025-986 77025-988 77025-990 77025-992 77025-994
Methyl-3-oxocyclobutanecarboxylate 98%
Methyl-3-oxocyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.127 g/mol
MDL Number: MFCD00604222
CAS Number: 695-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR