2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine 97%

Supplier: Ambeed
Warning

A114500-10G A114500-1G A114500-25G A114500-5G
77025-978EA 260.99 USD
77025-978 77025-980 77025-982 77025-984
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine 97%
2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂
MW: 189.0419 g/mol
MDL Number: MFCD09750204
CAS Number: 5466-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR