Ethyl 3-bromo-4-ethylbenzoate ≥95%

Supplier: Ambeed
Warning

A114343-250MG A114343-5G
76982-890EA 36.08 USD
76982-890 76982-892
Ethyl 3-bromo-4-ethylbenzoate ≥95%
Ethyl 3-bromo-4-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.12 g/mol
MDL Number: MFCD11112994
CAS Number: 1131615-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR