Benzyl (2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 96%

Supplier: Ambeed
Warning

A114274-1G A114274-250MG A114274-5G
77009-096EA 157.02 USD
77009-096 77009-098 77009-100
Benzyl (2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate 96%
Benzyl (2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₀H₂₃BClNO₄
MW: 387.67 g/mol
MDL Number: MFCD12546536
CAS Number: 1218791-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR