4,6-Dichloro-2-methylpyrimidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methyl-4,6-dichloropyrimidine
A114242-100G
A114242-10G
A114242-25G
A114242-500G
A114242-5G
76982-872EA
44.56
USD
76982-872
76982-874
76982-876
76982-878
76982-880
4,6-Dichloro-2-methylpyrimidine 97%
4,6-Dichloro-2-methylpyrimidine
Precaución: for Research Use Only
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol |
MDL Number:
MFCD00090472 CAS Number: 1780-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...