(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid 97%

Supplier: Ambeed
Danger

A114113-100MG A114113-10G A114113-1G A114113-250MG A114113-25G A114113-5G
76995-418EA 5 USD
76995-418 76995-420 76995-422 76995-424 76995-426 76995-428
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid 97%
(2R)-3-Bromo-2-hydroxy-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₄H₇BrO₃
MDL Number: MFCD09751022
CAS Number: 261904-39-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR