5-Bromo-2′-deoxycytidine 99%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

A114031-1G A114031-250MG A114031-25G A114031-5G
77025-934EA 57.29 USD
77025-934 77025-936 77025-938 77025-940
5-Bromo-2′-deoxycytidine 99%
5-Bromo-2′-deoxycytidine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN₃O₄
MW: 306.116 g/mol
MDL Number: MFCD00047496
CAS Number: 1022-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR