2-(Tritylthio)acetic acid 97%

Supplier: Ambeed
Warning

A114015-100G A114015-1G A114015-5G A114015-25G
77025-928EA 614.27 USD
77025-928 77025-930 77025-932 77549-218
2-(Tritylthio)acetic acid 97%
2-(Tritylthio)acetic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₂S
MW: 334.43 g/mol
MDL Number: MFCD00386069
CAS Number: 34914-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR