(1S,4R)-4-Aminocyclopent-2-enecarboxylic acid 95%

Supplier: Ambeed
Warning

A113951-1G A113951-250MG
76995-412EA 30.77 USD
76995-412 76995-414
(1S,4R)-4-Aminocyclopent-2-enecarboxylic acid 95%
(1S,4R)-4-Aminocyclopent-2-enecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MW: 127.14 g/mol
MDL Number: MFCD00211286
CAS Number: 134234-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR