3-Fluoro-2-hydroxypyridine 98%

Supplier: Ambeed
Warning

A113907-100G A113907-10G A113907-1G A113907-250MG A113907-25G A113907-5G
76995-400EA 245.01 USD
76995-400 76995-402 76995-404 76995-406 76995-408 76995-410
3-Fluoro-2-hydroxypyridine 98%
3-Fluoro-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄FNO
MDL Number: MFCD03095042
CAS Number: 1547-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR