Methyl 3-(p-tolyloxy)benzoate 95%

Supplier: Ambeed
Warning

A113894-1G A113894-250MG
77009-074EA 64.72 USD
77009-074 77009-076
Methyl 3-(p-tolyloxy)benzoate 95%
Methyl 3-(p-tolyloxy)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD06203838
CAS Number: 78303-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR