tert-Butyl 4-benzyl-3-oxopiperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A113779-100MG A113779-10G A113779-1G A113779-250MG A113779-5G
77025-914EA 49.87 USD
77025-914 77025-916 77025-918 77025-920 77025-922
tert-Butyl 4-benzyl-3-oxopiperazine-1-carboxylate 97%
tert-Butyl 4-benzyl-3-oxopiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₂N₂O₃
MW: 290.36 g/mol
MDL Number: MFCD09028511
CAS Number: 78551-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR