6-Methyl-4-phenylchroman-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-4-Phenylchromanone, 3,4-Dihydro-6-methyl-4-phenylcoumarin, 6-Methyl-4-phenyl-2-chromanone

A113698-10G A113698-1G A113698-25G A113698-5G A113698-250MG
77025-906EA 31 USD
77025-906 77025-908 77025-910 77025-912 77666-648
6-Methyl-4-phenylchroman-2-one 97%
6-Methyl-4-phenylchroman-2-one

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₂
MW: 238.2812 g/mol
MDL Number: MFCD03695019
CAS Number: 40546-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR