7-Bromo-4-fluorobenzo[b]furan 97%

Supplier: Ambeed
Warning

A113564-100MG A113564-250MG A113564-5G A113564-1G
77025-894EA 13 USD
77025-894 77025-896 77666-646 77871-464
7-Bromo-4-fluorobenzo[b]furan 97%
7-Bromo-4-fluorobenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₄BrFO
MDL Number: MFCD09056781
CAS Number: 253429-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR