1-Boc-6-fluoro-1H-indole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-6-Fluoro-1H-indole

A113487-100MG A113487-1G A113487-250MG A113487-5G
77025-880EA 15.92 USD
77025-880 77025-882 77025-884 77025-886
1-Boc-6-fluoro-1H-indole 98%
1-Boc-6-fluoro-1H-indole

Precaución: for Research Use Only

Formula: C₁₃H₁₄FNO₂
MW: 235.2542 g/mol
MDL Number: MFCD09909713
CAS Number: 1208459-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR