6-Chloro-3-methyluracil 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3-methylpyrimidine-2,4(1H,3H)-dione

A113466-100G A113466-10G A113466-25G A113466-500G A113466-5G
77009-058EA 30 USD
77009-058 77009-060 77009-062 77009-064 77009-066
6-Chloro-3-methyluracil 97%
6-Chloro-3-methyluracil

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
MDL Number: MFCD01074837
CAS Number: 4318-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR