5-Bromo-2-nitroisonicotinic acid 98%

Supplier: Ambeed
Warning

A113422-100MG A113422-1G A113422-250MG
76982-842EA 70.02 USD
76982-842 76982-844 76982-846
5-Bromo-2-nitroisonicotinic acid 98%
5-Bromo-2-nitroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrN₂O₄
MW: 247.00 g/mol
MDL Number: MFCD21648462
CAS Number: 1356412-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR