3,5-Dimethylisoxazole 95%

Supplier: Ambeed
Danger

A113298-10G A113298-25G A113298-5G
76995-348EA 7.43 USD
76995-348 76995-350 76995-352
3,5-Dimethylisoxazole 95%
3,5-Dimethylisoxazole

Precaución: for Research Use Only

Formula: C₅H₇NO
MW: 97.12 g/mol
MDL Number: MFCD00003156
CAS Number: 300-87-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR