N-Ethyltetrahydro-2H-pyran-4-amine 95%

Supplier: Ambeed
Warning

A113243-10G A113243-25G A113243-5G A113243-1G
77009-044EA 113.52 USD
77009-044 77009-046 77009-048 77570-730
N-Ethyltetrahydro-2H-pyran-4-amine 95%
N-Ethyltetrahydro-2H-pyran-4-amine

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MW: 129.20 g/mol
MDL Number: MFCD06658446
CAS Number: 211814-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR