(E)-Ethyl 4,4-dimethoxybut-2-enoate 95%

Supplier: Ambeed
Warning

A113235-10G A113235-1G A113235-250MG A113235-25G
76982-834EA 148.53 USD
76982-834 76982-836 76982-838 76982-840
(E)-Ethyl 4,4-dimethoxybut-2-enoate 95%
(E)-Ethyl 4,4-dimethoxybut-2-enoate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MW: 174.19 g/mol
MDL Number: MFCD06204472
CAS Number: 114736-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR