5-Methyl-1H-indazole-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A113115-1G
A113115-250MG
A113115-5G
77009-030EA
44.56
USD
77009-030
77009-032
77009-034
5-Methyl-1H-indazole-3-carboxylic acid 95%
5-Methyl-1H-indazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C9H8N2O2 MW: 176.17 g/mol |
MDL Number:
MFCD05663986 CAS Number: 1201-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...