1-Amino-5-bromoisoquinoline 97%

Supplier: Ambeed
Warning

A113105-250MG A113105-1G A113105-100MG
77025-858EA 44 USD
77025-858 77570-724 77871-460
1-Amino-5-bromoisoquinoline 97%
1-Amino-5-bromoisoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD11226920
CAS Number: 852570-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR