5-(Benzyloxy)-2-(hydroxymethyl)-4H-pyran-4-one 97%

Supplier: Ambeed
Warning

A113052-1G A113052-250MG A113052-25G A113052-5G
76995-330EA 48.81 USD
76995-330 76995-332 76995-334 76995-336
5-(Benzyloxy)-2-(hydroxymethyl)-4H-pyran-4-one 97%
5-(Benzyloxy)-2-(hydroxymethyl)-4H-pyran-4-one

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₄
MDL Number: MFCD01927596
CAS Number: 15771-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR