tert-Butyl 4-(4-carbamoylphenoxy)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A112976-100MG A112976-1G A112976-250MG
77009-024EA 21 USD
77009-024 77009-026 77009-028
tert-Butyl 4-(4-carbamoylphenoxy)piperidine-1-carboxylate 97%
tert-Butyl 4-(4-carbamoylphenoxy)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₄
MDL Number: MFCD24470001
CAS Number: 609781-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR