PX 12 98%

Supplier: Ambeed
Warning

A112791-10MG A112791-1MG A112791-25MG A112791-5MG
77008-994EA 63 USD
77008-994 77008-996 77008-998 77009-000
PX 12 98%
PX 12

Precaución: for Research Use Only

Formula: C₇H₁₂N₂S₂
MDL Number: MFCD18086851
CAS Number: 141400-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR