1-(2-Aminophenyl)ethanol 98%

Supplier: Ambeed
Warning

A112768-1G A112768-25G A112768-5G
76995-310EA 32.89 USD
76995-310 76995-312 76995-314
1-(2-Aminophenyl)ethanol 98%
1-(2-Aminophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.17 g/mol
MDL Number: MFCD12198838
CAS Number: 10517-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR