5-Chloro-3-methyl-2-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
A112571-100MG
A112571-250MG
A112571-5G
A112571-1G
77008-984EA
22.28
USD
77008-984
77008-986
77570-692
77683-030
5-Chloro-3-methyl-2-nitropyridine 98%
5-Chloro-3-methyl-2-nitropyridine
Precaución: For research use only.
Formula:
C₆H₅ClN₂O₂ MW: 172.57 g/mol |
MDL Number:
MFCD18260751 CAS Number: 1211532-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...