5-Bromo-1,4-dimethylimidazole 97%

Supplier: Ambeed
Warning

A112508-100MG A112508-250MG
76982-800EA 120 USD
76982-800 76982-802
5-Bromo-1,4-dimethylimidazole 97%
5-Bromo-1,4-dimethylimidazole

Precaución: for Research Use Only

Formula: C₅H₇BrN₂
MDL Number: MFCD23378484
CAS Number: 861325-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR