(1-Chloroisoquinolin-4-yl)boronic acid 97%

Supplier: Ambeed
Warning

A112457-100MG A112457-1G A112457-250MG A112457-5G
76982-796EA 54 USD
76982-796 76982-798 77885-046 77885-047
(1-Chloroisoquinolin-4-yl)boronic acid 97%
(1-Chloroisoquinolin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₇BClNO₂
MDL Number: MFCD09959721
CAS Number: 848841-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR