Ethyl 4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidine-5-carboxylate 98%

Supplier: Ambeed
Warning

A112271-1G A112271-250MG
76982-786EA 190.97 USD
76982-786 76982-788
Ethyl 4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidine-5-carboxylate 98%
Ethyl 4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₂FN₃O₄S
MW: 395.45 g/mol
MDL Number: MFCD12828196
CAS Number: 147118-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR