7-Isoquinolinecarboxylic acid 95%

Supplier: Ambeed
Warning

A112196-250MG A112196-1G A112196-100MG
77025-814EA 42 USD
77025-814 77570-676 77599-616
7-Isoquinolinecarboxylic acid 95%
7-Isoquinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MDL Number: MFCD10699235
CAS Number: 221050-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR