1-Bromo-3-isobutylbenzene 98%

Supplier: Ambeed
Warning

A112166-100MG A112166-250MG
77008-976EA 55 USD
77008-976 77008-978
1-Bromo-3-isobutylbenzene 98%
1-Bromo-3-isobutylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br
MDL Number: MFCD26405543
CAS Number: 139155-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR