4,4'-Oxybis(benzoic acid) 97%

Supplier: Ambeed
Warning

Synonyms: 4,4′-Oxybis(benzoic acid)

A112150-100G A112150-10G A112150-25G A112150-500G
76995-282EA 29.71 USD
76995-282 76995-284 76995-286 77565-964
4,4'-Oxybis(benzoic acid) 97%
4,4'-Oxybis(benzoic acid)

Precaución: For research use only.

Formula: C₁₄H₁₀O₅
MW: 258.23 g/mol
MDL Number: MFCD00013988
CAS Number: 2215-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR