O-[2-Oxo-1(2H)-pyridyl]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%

Supplier: Ambeed
Danger

Synonyms: O-(2-Oxo-1(2H)pyridyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, O-(1,2-Dihydro-2-oxo-1-pyridyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TPTU

A112125-10G A112125-1G A112125-25G A112125-5G
76995-274EA 23 USD
76995-274 76995-276 76995-278 76995-280
O-[2-Oxo-1(2H)-pyridyl]-N,N,N',N'-tetramethyluronium tetrafluoroborate 97%
O-[2-Oxo-1(2H)-pyridyl]-N,N,N',N'-tetramethyluronium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₁₆BF₄N₃O₂
MW: 297.06 g/mol
MDL Number: MFCD00075475
CAS Number: 125700-71-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR